CC2695P Series Dual Wire Coil Cords, Transforming Technologies
Supplier: Transforming Technologies
CC2695P-10 CC2695P-20 CC2695P-5
76489-580EA 38.46 CAD
76489-580 76489-582 76489-584
CC2695P Series Dual Wire Coil Cords, Transforming Technologies
Grounders Grounding Wires
Polyurethane coil insulation and strain relief molding offers excellent coil memory and endurance.
  • 'One snap' dual wire design contains two snaps in a molded head
  • Innovative parking jack to hold coil cord plug
  • 5, 10, and 20' lengths (extended)
  • 1 meg ohm resistor
The molded snap head provides a convenient parking jack for the cord when not in use. Two female snaps, a 3.5 mm phono plug, and 1 meg resistor are standard.
Order Now
SPECIFICATIONS
Environmentally Preferable