extraktLAB DrainDroyd Filtration System, United Science
Supplier: United Science
70-3009 70-3010
CA76501-188EA 22397 CAD
CA76501-188 CA76501-190
extraktLAB DrainDroyd Filtration System, United Science
Funnels Filtration Funnels Büchner Funnels
DrainDroyd filtration system is designed for rapid filtration of solids from liquids. Standard DrainDroyd base compatible with Octolid, terpene condenser, filter and jacketed filter top accessories.
  • High throughput filtration
  • Containment vessel for product isolation
  • GMP-compliant stainless-steel construction
  • Superb cleanability
  • Starter kit includes everything needed
  • Multiple accessories to expand methods
  • Adjustable casters
The DrainDroyd filtration system makes high-throughput wax removal from winterized ethanol solutions. It eliminates theMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable