extraktLAB MPCart Pump, United Science
Supplier: United Science
10-0020
CA76501-180EA 50902.25 CAD
CA76501-180
extraktLAB MPCart Pump, United Science
Monitoring Systems CO2 and Temperature Monitoring Systems
Step-up pump for filling the CO₂ cage from a bulk cryogenic source. Features a 20 lbs./min. fill rate, touch screen, 4 to 20 mAmp connectivity to CO₂ cage, and 100' of flexible discharge hose. May be permanently plumbed in line.
  • Mobile or permanently plumbed inline CO₂ pump solution
  • Transports liquid CO₂ more than 100 feet
  • Integrates with CO₂ cage
  • CO₂ cage automatically signals to turn pump on
  • 1500 psi max. pressure
The MPCart is a step-up CO₂ pump mounted on a moveable cart that is integrated with the operating functions of the CO₂ cage. It draws CO₂ from outdoor bulk storage tanks at 300 psi, and steps the pressure up to 800 psi to keep the CO₂ cage full.
The MPCart is controlled by the electronics and touch screen display onboard the CO₂ cage for automated replenishment. It comes with 120 feet of CO₂ hose and power cabling that connects to the CO₂ cage for maximum versatility in factory floor placement.
Certifications: UL/CSA/ISO/ASME.
Ordering information: Two-year warranty.