Sole Grounders, 2 Meg, Transforming Technologies
Supplier: Transforming Technologies
HG6012 HG6022 HG6032 HG6052
76489-734EA 24.55 CAD
76489-734 76489-736 76489-738 76489-740
Sole Grounders, 2 Meg, Transforming Technologies
Grounders Heel and Toe Grounders
Full coverage sole grounders provide 20 times the surface contact of heel grounders.
  • Wider and more consistent contact area ensures grounding
  • Rapid and complete static dissipation due to constant contact through entire walking motion
ESD Sole Grounders (or ESD Foot Grounders) offer over 20 times the surface contact of standard ESD heel straps. This surface area advantage extends the life of the sole grounder over standard heel grounders, which fail over time due to even the slightest contamination, necessitating replacement or cleaning, wastingMore Product Information