HBIO ECM 630 Electroporation System, BTX
Supplier: Harvard Apparatus
45-0658 47-0040 47-0043 47-0045 47-0050 47-0060 47-0070 47-0080 47-0086
76324-956EA 25632.92 CAD
76324-956 76324-958 76324-972 76324-984 76324-986 76324-988 76324-990 76324-992 76324-994 76324-996
HBIO ECM 630 Electroporation System, BTX
Electroporation Systems Electroporation Kits
Our electroporation systems and accessories are primarily used for bacteria and yeast transformation. High throughput (25- and 96-well) models are available.
  • High throughput 25- and 96-well electroporation
  • Voltage range of 5 to 3000 V
  • Capacitance range of 25 to 3275 µF in 25 µF steps in LV mode; 10, 25, 35, 50, 60, 75, 85 µF in HV mode
  • Large, easy to use color touchscreen interface
Wide and accurate voltage, capacitance, and internal resistance settings; Preset protocols for common bacteria, yeast, and microorganism types; Unlimited ability to add andMore Product Information