Wolfgang Warmbier PGT120 Tester with RS232 Interface, Transforming Technologies
Supplier: Transforming Technologies
7100.PGT120.D2 7100.PGT120.D3 7100.PGT120.DT PGT120.COM.DT3 PGT120.COMSOFT PGT120.COMTC18 PGT120.COM.US PGT120.CU.12 7100.PGT120.SE 7100.PGT120.SM PGT120.TEST.10 PGT120.TEST.11 PGT120.TEST.12 PGT120.TEST.13 PGT120.TEST.15 PGT120.TEST.16 PGT120.TEST.17 PGT120.XK.V3
76489-530EA 391.57 CAD
76489-530 76489-532 76489-534 76489-536 76489-538 76489-540 76489-542 76489-544 76489-546 76489-548 76489-550 76489-552 76489-554 76489-556 76489-558 76489-560 76489-562 76489-566
Wolfgang Warmbier PGT120 Tester with RS232 Interface, Transforming Technologies
Static Control Monitors
The PGT120.COM ESD grounding tester takes quick and accurate resistance measurements of ESD wrist straps, ESD footwear, heel grounders, and smocks to store test data on a computer.
  • Independently test the functioning of ESD footwear and wrist strap grounding products and signals
  • 'Smart' ESD combo tester integrates with a computer to record test results into a database
  • Can work with other USB devices such as label printer, RFID reader, card reader, keypad, light signal, bar code scanner, and more
  • Automatically test footwear in hands-free mode
The PGT120.COM personnel grounding tester takes quick and accurate resistance measurements of the ESD grounding products, and features the ability to integrate with a computer to store test data. The unit operates with three independent measuring circuits for the left shoe, the right shoe, and the wrist strap. The shoes can even be measured in a hands-free mode that allows staff members to be tested automatically when standing on the plate. The PGT120.COM can be connected to an automatic door opening system which allows entry with only passing results.
The unit is supplied complete with footwear testing electrode, flexible cord, transformer with power cord, and calibration protocol. Recommended calibration interval is 24 months. Corresponding wall mounting plate, and electrode anti-fatigue mat are sold separately.