Chlorate (ClO₃)- Single-Element Ion Anion Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Supplier: VWR International
BDH0815-125ML
BDH0815-500ML
BDH0815-125MLEA
205.42
CAD
BDH0815-125ML
BDH0815-500ML
Chlorate (ClO₃)- Single-Element Ion Anion Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Standards
Anionic Standards
Chlorate Standards
Ion chromatography & ion selective electrode (ISE) standard.
- Single and multi-element standards for ion chromatography available
- Stable and traceable