Bromide (Br-) Single-Element Ion Anion Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Supplier: VWR International
BDH0811-125ML
BDH0811-500ML
BDH0811-125MLEA
132.64
CAD
BDH0811-125ML
BDH0811-500ML
Bromide (Br-) Single-Element Ion Anion Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Standards
Anionic Standards
Bromide Standards
Ion chromatography & ion selective electrode (ISE) standard.
- Single and Multi-Element Standards for Ion Chromatography available
- Stable and traceable