Cove N Curve™ Mop Adapter, Micronova
Supplier: Micronova
QPRCVT-16 QPRCVT-09B
76407-418EA 201.78 CAD
76407-418 76407-420
Cove N Curve™ Mop Adapter, Micronova
Mops
This uniquely shaped mop adapter transforms a traditional flat mop design into a curved cleaning surface. Designed to address coved joints, exterior of pipes, the interior of drums and small tanks, as well as other curved surfaces.
  • Constructed from PTFE with a Polyketone swivel joint
  • Swivel joint allows an easy glide
  • ACME thread adapter
  • Resistant to common cleaning chemicals and disinfectants
  • Available in 9" or 16" (23 or 41 cm) lengths
  • Autoclavable
The Micronova Cove N Curve™ Mop Adapter is designed for use with SnapMop™ and PocketMop™ series flatMore Product Information