NovaMop™ Self Wringing Mops, Micronova
Supplier: Micronova
QPNMSW-18 QPNMSW-14
75856-886EA 580.48 CAD
75856-886 75856-892
NovaMop™ Self Wringing Mops, Micronova
Mops
A flat head mop featuring a unique self-wringing mechanism that folds the mop head in half and draws upward for uniform wringing, every time. The 360° swivel joint allows full rotation of the mop head for use on floors, walls, and ceilings in the cleanroom.
  • Self wringing mop adapter with 360° swivel
  • One-step wringing process
  • Ideal, uniform wringing pressure
  • ACME thread adapter
  • Resistant to common cleaning chemicals and disinfectants
  • Autoclavable
The Micronova NovaMop™ Self Wringing Mop is designed for use with M-Zone® FlatMop™, SnapMop™, and PocketMop™ series flat head mops and connects directly to the STA series threaded handles, fixed length or extendible. Can also be used with SSU series stainless steel handles, with the addition of the SSA-3 adapter.
Accessories information: All components are ordered separately. For additional handle lengths, see Universal Mop Handles or Plastic Mop Handles from Micronova.