SlipGuard Impervious Shoe Covers, Apex Aseptic Products
Supplier: APEX ASEPTIC PRODUCTS, LLC.
2680WHS/M 2680WHL/XL 2680WH2XL 2680SBS/M 2680SBL/XL 2680SB2XL
76488-362CS 121.88 CAD
76488-362 76488-364 76488-366 76488-368 76488-370 76488-372
SlipGuard Impervious Shoe Covers, Apex Aseptic Products
Disposable Garments
SlipGuard Impervious Shoe Covers are made of durable, skid-resistant PE coated non-woven fabrics. These garments are impervious and lint-free.
  • Durable, tear-resistant, water-resistant
  • Anti-skid treated to help prevent accidental falls
  • Latex-free, Silicone-free, and nonsterile
  • Low particle shedding, category 1 per Helmke Drum Test
  • Manufactured in controlled environment
  • Reasonable cost, affordable for everyday use
SlipGuard Impervious Shoe Covers are ideal for use in pharmaceutical manufacturing facilities, animal labs, research centers, medical establishments, and cleanroom environments.
Certifications: Manufacturing facility is ISO 9001 and ISO 13485 certified. FDA registered and inspected.
Packaging: Shoe covers are packaged 50 per bag, 300 per case.