Non-Woven Surgical Hood, Apex Aseptic Products
Supplier: APEX ASEPTIC PRODUCTS, LLC.
2050SMS-B 2050PP-WH
76488-374CS 163.58 CAD
76488-374 76488-376
Non-Woven Surgical Hood, Apex Aseptic Products
Disposable Garments
Non-woven surgical hoods provide splash protection for the entire head and neck, and prevent loose hair from falling into the work environment.
  • Crown hood, with elastics around face
  • Low particle shedding, rated category 1 per Helmke Drum Test
  • Latex-free, TSE-free, BSE-free, and non-sterile
  • Manufactured in controlled environment
Splash-resistant and breathable hoods are made with lightweight SMS fabrics.
Certifications: Manufacturing facility is ISO 9001 and ISO 13485 certified. FDA registered and inspected.
Packaging: Hoods are packaged 100 per bag, 500 per case.