Menda ESD Safe Trigger Spray Bottles, Desco
Supplier: Menda
35796 35797 35798 35799
76490-398EA 20.37 CAD
76490-398 76490-400 76490-402 76490-404
Menda ESD Safe Trigger Spray Bottles, Desco
Bottles Spray Bottles
Menda ESD Safe trigger spray bottles provide controlled delivery of solvents, cleaners, water, oils, or other compatible liquids at the ESD workstation.
  • Low Density Polyethylene (LDPE) material
  • Long lasting dissipative properties
  • Rtt: 1×10⁴ to <1×10¹¹ ohms tested per ANSI/ESD STM11.12
  • Trigger spray nozzle features spray, stream, and off settings
  • Trigger spray head is insulative
  • No migratory additives; reduces the chance for contamination from the bottle
  • Embossed with ESD protective symbol
  • Made in the United States of America
These LDPE Trigger spray bottles are embedded with long lasting dissipative properties and are available in blue or yellow, in 8 or 16 ounce sizes. Menda offers non-printed options as well as custom printing capabilities.