Menda ESD Safe Wash Bottles, Desco
Supplier: Menda
35790 35791 35792 35793 35794 35795
76490-386EA 19.1 CAD
76490-386 76490-388 76490-390 76490-392 76490-394 76490-396
Menda ESD Safe Wash Bottles, Desco
Bottles Wash Bottles
Menda ESD Safe wash bottles with a swan neck spout are used to hold water for wetting sponges at soldering stations, and to hold cleaning agents or similar liquids.
  • Low Density Polyethylene (LDPE) durAstatic® Wash bottles with swan neck spout
  • Long lasting dissipative properties
  • Rtt: 1×10⁴ to <1×10¹¹ ohms tested per ANSI/ESD STM11.12
  • No migratory additives; reduces the chance for contamination from the bottle
  • Embossed with ESD protective symbol
  • Made in the United States of America
These LDPE wash bottles are embedded with long lasting dissipativeMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable