Menda ESD Safe Vinyl Breakaway Lanyard, Desco
Supplier: Desco Industries
35108
76490-384EA 9.2 CAD
76490-384
Menda ESD Safe Vinyl Breakaway Lanyard, Desco
Personal Identification ID Card Holders
Ideal for use by electronic manufacturers working with ESD sensitive components. Strap snaps around the neck and attaches to clip-on or retractable badge holder.
  • Dissipative vinyl 1×10⁴ to <1×10¹¹ ohms, per ANSI/ESD STM11.11
  • Designed for short term use
  • Snaps are ³/₈" (10 mm) in diameter
  • Made with one layer of 0.020" (0.5 mm) matte soft vinyl, in between two layers of 0.012" (0.3 mm) thick ESD vinyl
  • Soft, frosty clear vinyl presents a low profile that doesn’t compete with clothing
  • With a nominal 15 pound force applied to lanyard it will break
The Menda ESD Safe Vinyl Breakaway Lanyard is ideal for use by electronic manufacturers working with ESD sensitive components. Strap snaps around the neck and attaches to clip-on or retractable badge holder. Meant for short term use and for additional safety features a nominal 15 pound force breaking point.
Certifications: Meets ANSI/ESD S20.20 and ANSI/ESD S541.