Non-Woven Protective Gowns, Apex Aseptic Products
Supplier: APEX ASEPTIC PRODUCTS, LLC.
3360B 3360P
76488-406CS 146.86 CAD
76488-406 76488-408 76488-410
Non-Woven Protective Gowns, Apex Aseptic Products
Disposable Garments
Non-Woven Protective Gowns are fluid resistant and breathable.
  • Fluid resistant, breathable, and economical
  • Latex-free, TSE-free, BSE-free
  • Low particle shedding, rated category 1 per Helmke Drum Test
  • Manufactured in controlled environment
The entire gown is made with medium weight non-woven fabrics. These knee length gowns feature a full back, knit cuffs, and back ties that allow for adjustable sizing.
Certifications: Manufacturing facility is ISO 9001 and ISO 13485 certified. FDA registered and inspected.
Packaging: Gowns are packaged 10 perMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Gender
Garment application