Core Samplers, Burkle
Supplier: BURKLE INC
5310-0001 5310-0002 5310-0003
76501-314EA 1423.03 CAD
76501-314 76501-316 76501-318
Core Samplers, Burkle
Samplers Solid Samplers
The core sampler can be used to take samples of bulk materials, which can easily become stuck or settle during sampling due to their particle form.
  • Stainless steel AISI 316 construction
  • For bulk materials that become stuck or settle easily
  • Order includes core sampler (consisting of boring rod and boring tube), fixing clip
In comparison to standard all-layer samplers for bulk materials, the two-part core sampler has no gap dimensions in which the medium can become stuck while it is being removed. This makes it possible to take a proper sample of powders and granulates through multiple layers, for example glass balls or metal powders.
Depending on the size, the stainless steel sampler can be used for a wide range of different containers, such as large bags or small containers. The bulk material sampler takes a representative core sample and is ideal for use in various laboratory and industrial applications.