Extra-Safe Lab Coats with Three Pockets, Apex Aseptic Products
Supplier: APEX ASEPTIC PRODUCTS, LLC.
3660WHXS-T 3660WHS-T 3660WHM-T 3660WHL-T 3660WHXL-T 3660WH2XL-T 3660WH3XL-T 3660WH4XL-T 3660WH5XL-T 3660MBS 3660MBM 3660MBL 3660MBXL 3660MB2XL 3660MB3XL 3660MB4XL 3660MB5XL 3660RYS 3660RYM 3660RYL 3660RYXL 3660BBXS 3660BBS 3660BBM 3660BBL 3660BBXL 3660BB2XL 3660BB3XL 3660BB4XL 3660BB5XL 3660BKXS 3660BKS 3660BKM 3660BKL 3660BKXL 3660BK2XL 3660BK3XL 3660BK4XL 3660BK5XL 3660CBXS 3660CBS 3660CBM 3660CBL 3660CBXL 3660CB2XL 3660CB3XL 3660CB4XL 3660CB5XL 3660WHXS-K 3660WHS-K 3660WHM-K 3660WHL-K 3660WHXL-K 3660WH2XL-K 3660WH3XL-K 3660WH4XL-K 3660WHX5XL-K 3660SBXS 3660SBS 3660SBM 3660SBL 3660SBXL 3660SB2XL 3660LPXS 3660LPS 3660LPM 3660LPL 3660LPXL 3660LP2XL 3660LP3XL 3660RBXS 3660RBS 3660RBM 3660RBL 3660RBXL 3660RB2XL 3660RB3XL 3660TEXS 3660TES 3660TEM 3660TEL 3660TEXL 3660TE2XL 3660TE3XL 3660TE4XL 3660TE5XL 3660PPXS 3660PPS 3660PPM 3660PPL 3660PPXL 3660PP2XL 3660PP3XL 3660PP4XL 3660PP5XL 3660CYXS 3660CYS 3660CYM 3660CYL 3660CYXL 3660CY2XL 3660CY3XL 3660CY4XL 3660CY5XL
76488-492CS 231.74 CAD
76488-492 76488-494 76488-496 76488-498 76488-500 76488-502 76488-504 76488-506 76488-508 76488-510 76488-512 76488-514 76488-516 76488-518 76488-520 76488-522 76488-524 76488-526 76488-528 76488-530 76488-532 76488-534 76488-536 76488-538 76488-540 76488-542 76488-544 76488-546 76488-548 76488-550 76488-552 76488-554 76488-556 76488-558 76488-560 76488-562 76488-564 76488-566 76488-568 76488-570 76488-572 76488-574 76488-576 76488-578 76488-580 76488-582 76488-584 76488-586 76488-588 76488-590 76488-592 76488-594 76488-596 76488-598 76488-600 76488-602 76488-604 76488-606 76488-608 76488-610 76488-612 76488-614 76488-616 76488-618 76488-620 76488-622 76488-624 76488-626 76488-628 76488-630 76488-632 76488-634 76488-636 76488-638 76488-640 76488-642 76488-644 76488-646 76488-648 76488-650 76488-652 76488-654 76488-656 76488-658 76488-660 76488-662 76488-664 76488-666 76488-668 76488-670 76488-672 76488-674 76488-676 76488-678 76488-680 76488-682 76488-684 76488-686 76488-688 76488-690 76488-692 76488-694 76488-696 76488-698
Extra-Safe Lab Coats with Three Pockets, Apex Aseptic Products
Disposable Garments
These Extra-Safe lab coats are highly fluid resistant, yet breathable and comfortable to wear.
  • Made with SMS fabrics
  • Hydro-Head Fluid Resistant Rating: 529 mm
  • Spray Impact Rating: 0.53 g
  • Easy breathability, latex-free, TSE-free, BSE-free
  • Knee length, snap front, three pockets, knit cuffs
  • Manufactured in controlled environment
  • Low particle shedding, rated Category 1 per Helmke Drum Test
  • Autoclavable up to five times for reuse if desired
Extra-Safe lab coats are available in 12 attractive color choices to brighten up any operation or to color-codeMore Product Information