LaMotte® Dissolved Oxygen Water Test Kit (Winkler Method)
Supplier: LA MOTTE CHEMICAL PROD CO SE
5860-CN-01
470335-420EA 123.25 CAD
470335-420
LaMotte® Dissolved Oxygen Water Test Kit (Winkler Method)
Environmental Test Kits
Precalibrated test tubes and labware give fast, accurate results on-site, in the classroom, or in the lab.
  • Range: 0 to 10 ppm
  • Materials for 50 tests
Reagents, labware, and step-by-step instructions are furnished in a compact, waterproof carrying case.