Vacuum Fluids for Rotary Vane Pumps, Agilent Technologies
Supplier: Agilent
CrossLab
X3760-64004 X3760-64004
CAX3760-64004EA 218.25 CAD
AGX3760-64004 CAX3760-64004
Vacuum Fluids for Rotary Vane Pumps, Agilent Technologies
Lubricants
Agilent pump fluids offer the best performance for your DS-and MS-series pumps.
  • Agilent vacuum pump fluids are specifically formulated to provide optimal performance for each pump design and capability
  • The DS-19 Oil is an extremely-low vapor pressure fluid for use in dual stage pumps to allow the pumps to achieve the lowest possible base pressure while providing the lubricity and wear resistance required for the longest possible pump life
As different vacuum pumps are applied for different uses, vacuum pump oils are tailored to different pump operations. Appropriate vapor pressure, viscosity, resistance to heat or chemical attack, and other factors distinguish which pump fluids provide for optimal performance. Agilent vacuum pump fluids have been specifically selected for each Agilent pump model, assuring that the pumps can fully achieve their operating specifications.