CrossLab Torches for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G3280-80053 G3280-80080 G3270-80043 G3280-80081
CAG3280-80053EA 1013.72 CAD
CAG3280-80053 CAG3280-80080 CAAGG327080043 CAAGG328080081
CrossLab Torches for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
One-piece quartz torches for Agilent ICP-MS, including standard and torches for organic solvents.
  • The standard ICP-MS torch is an easy-mount, one-piece quartz torch with 2.5 mm int.Ø injector, to ensure a robust plasma that efficiently decompose the sample matrix, minimizing matrix deposition, signal drift and routine maintenance
  • The torch postitioning is maintained by the mounting bracket, ensuring consistent repositioning following routine maintenance or when switching torches
  • The torch is then auto-aligned by the motorized torch box with software
More Product Information