CrossLab Torch Bonnets, Shield Plates and Components for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G1833-65419 G1833-65421
CA76515-248EA 2842.95 CAD
CA76515-248 CA76515-250
CrossLab Torch Bonnets, Shield Plates and Components for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Long life platinum shield-plates and quartz bonnets for Agilent ICP-MS Torches.
  • Shield torch system (STS) comprises a platinum shield, and a quartz bonnet, positioned between the torch and the work coil
  • Replacement platinum shields and quartz bonnets are availble for all 7700, 7800, 7900 and 8800, 8900 ICP-MS
  • Long life platinum shield-plates and quartz bonnets should be inspected and replaced when there is any sign of physical damage such as cracks or chips
The Agilent plasma and shield torch system (STS) provides precise ion energy control, ensuringMore Product Information