1,3,4-tri-O-Acetyl-2-azido-2,6-dideoxy-L-talose ≥95%

Supplier: AFG Bioscience

C51793-25MG C51793-50MG
76619-706EA 791.2 CAD
76619-706 76619-708
1,3,4-tri-O-Acetyl-2-azido-2,6-dideoxy-L-talose ≥95%
1,3,4-tri-O-Acetyl-2-azido-2,6-dideoxy-L-talose

Order Now

Specification Test Results

Purity Limit >95%
Appearance white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR