2-Azido-2,6-dideoxy-L-talose ≥95%

Supplier: AFG Bioscience

C51792-10MG C51792-25MG C51792-50MG
76619-700EA 791.2 CAD
76619-700 76619-702 76619-704
2-Azido-2,6-dideoxy-L-talose ≥95%
2-Azido-2,6-dideoxy-L-talose

Order Now

Specification Test Results

Purity Limit >95%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR