Lewis X tetrasaccharide-PEG24-azide ≥95%

Supplier: AFG Bioscience

C51779-1MG C51779-0.1MG C51779-0.25MG
76619-678EA 3146.24 CAD
76619-678 77888-813 77888-814
Lewis X tetrasaccharide-PEG24-azide ≥95%
Lewis X tetrasaccharide-PEG24-azide

Order Now

Specification Test Results

Purity Limit >95%
Appearance White Waxy Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR