Mannose-α-D-PEG24-N3 ≥95%

Supplier: AFG Bioscience

C51777-1MG C51777-5MG
76619-674EA 395.6 CAD
76619-674 76619-676
Mannose-α-D-PEG24-N3 ≥95%
Mannose-α-D-PEG24-N3

Order Now

Specification Test Results

Purity Limit >95%
Appearance White Waxy Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR