2-Azido-2-deoxy-L-fucose ≥95%
Supplier: AFG Bioscience
C51551-25MG
C51551-50MG
76619-566EA
700.94
CAD
76619-566
76619-568
2-Azido-2-deoxy-L-fucose ≥95%
2-Azido-2-deoxy-L-fucose
Specification Test Results
Purity Limit | >95% |
Appearance | white powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...