Fmoc-L-Tyr-(β-D-Gal(Ac)4)-OH ≥95%
Supplier: AFG Bioscience
Synonyms:
N-α-(9-Fluorenylmethyloxycarbonyl)-O-β-(2,3,4,6-tetra-O-acetyl-β-D-galactopyranosyl)-L-tyrosine
C51082-100MG
C51082-250MG
C51082-25MG
76619-370EA
1183.09
CAD
76619-370
76619-372
76619-368
Fmoc-L-Tyr-(β-D-Gal(Ac)4)-OH ≥95%
Fmoc-L-Tyr-(β-D-Gal(Ac)4)-OH
Specification Test Results
Purity Limit | >95% |
Appearance | White powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...