D-Fructose 1-phosphate dipotassium salt 95%

Supplier: AFG Bioscience

C50707-10MG C50707-5MG
76619-152EA 1582.4 CAD
76619-152 76619-150
D-Fructose 1-phosphate dipotassium salt 95%
D-Fructose 1-phosphate dipotassium salt

Caution: Store at -20°C

Order Now

Specification Test Results

Purity Limit >95%
Appearance White Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR