HyClone™ Donor Equine Serum, U.S. origin, HyClone products (Cytiva)
Supplier: Cytiva
3007403IH25-40 3007403IR25-40 3007404IR25-40 3007404IR30-45 SH30074.02HI SH30074.04
CA76534-628EA 151.77 CAD
CA76534-628 CA76324-874 CA76534-632 CA76534-630 CA82013-588 CA76236-290
HyClone™ Donor Equine Serum, U.S. origin, HyClone products (Cytiva)
Sera (Serum)
The animals are fed an enriched diet to ensure proper nutrition and are fasted prior to bleeding to reduce lipid concentration. The horses are bled in an enclosed facility that is specially equipped for aseptic collection. Coggins tests for equine infectious anemia are routinely performed on each horse.
  • Tested for equine infectious anemia
  • Complete traceability back to original source
  • Virus panel testing according to 9 CFR 113.53
onor Equine Serum is filtered through three sequential 100 nm (0,1 μm) pore size-rated filters. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.