Guanidinium hydrochloride solution 8 M, Pierce™

Supplier: Thermo Scientific

24115
CAPI24115EA 712.44 CAD
CAPI24115
Guanidinium hydrochloride solution 8 M, Pierce™
Guanidine hydrochloride

Thermo Scientific Pierce 8M Guanidine-HCl Solution is an accurately formulated, ready-to-use solution of guanidine hydrochloride, that can be easily diluted and pH-adjusted to any concentration below 8M.


  • Any concentration below 8M guanidine can be quickly and easily prepared.
  • No wasting valuable research time weighing and dissolving guanidine crystals
  • Free from UV absorbing materials in the range of 225-300nm
  • Free from heavy metal contaminants
  • Free from degradation products such as ammonia
  • Excellent stability


Applications of Guanidine Hydrochloride:
• Solubilizing proteins from inclusion bodies
• Increasing solubility of hydrophobic peptides and proteins
• Denaturing proteins

Formula: NH₂C(=NH)NH₂·HCl
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR