Dithiothreitol (DTT, Cleland's reagent), Pierce™

Supplier: Thermo Scientific

Synonyms: DL-1,4-DithiothreitolDTTcis-2,3-Dihydroxy-1,4-butanedithiol(R*,R*)-1,4-Dimercapto-2,3-butandiol

20290 20290
CAPI20290EA 468.36 CAD
CAPI20290 PI20290
Dithiothreitol (DTT, Cleland's reagent), Pierce™
Dithiothreitol (DTT, Cleland's reagent)

Dithiothreitol is a popular reducing agent for many protein research applications, including reducing polyacrylamide electrophoresis (SDS-PAGE).


  • Dithiothreitol, Cleland's Reagent Reduces Disulfide Bonds

Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00004877
CAS Number: 3483-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR