WHEATON® CompletePAK® Sterile Vial Kits, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W991000-CP W991001-CP W991002-CP W991003-CP W991004-CP W991005-CP W991006-CP W991007-CP W991008-CP W991009-CP W991010-CP W991011-CP W991012-CP W991013-CP W991014-CP W991015-CP W991016-CP W991017-CP W991000-CPA W991001-CPA W991002-CPA W991003-CPA W991004-CPA W991005-CPA W991006-CPA W991007-CPA W991008-CPA W991009-CPA W991010-CPA W991011-CPA W991012-CPA W991013-CPA W991014-CPA W991015-CPA W991016-CPA W991017-CPA W991000-CPMP W991001-CPMP W991002-CPMP W991003-CPMP W991004-CPMP W991005-CPMP W991006-CPMP W991007-CPMP W991008-CPMP W991009-CPMP W991010-CPMP W991011-CPMP W991012-CPMP W991013-CPMP W991014-CPMP W991015-CPMP W991016-CPMP W991017-CPMP W991000A-CPMP W991001A-CPMP W991002A-CPMP W991003A-CPMP W991004A-CPMP W991005A-CPMP W991006A-CPMP W991007A-CPMP W991008A-CPMP W991009A-CPMP W991010A-CPMP W991011A-CPMP W991012A-CPMP W991013A-CPMP W991014A-CPMP W991015A-CPMP W991016A-CPMP W991017A-CPMP W987000-S W987001-S W987002-S W987000A-S W987001A-S W987002A-S
76489-350CS 2788.81 CAD
76489-350 76489-352 76489-354 76489-356 76489-358 76489-360 76489-362 76489-364 76489-366 76489-368 76489-370 76489-372 76489-374 76489-376 76489-378 76489-380 76489-382 76489-384 76489-386 76489-388 76489-390 76489-392 76489-394 76489-396 76489-398 76489-400 76489-402 76489-404 76489-406 76489-408 76489-410 76489-412 76489-414 76489-416 76489-418 76489-420 76489-422 76489-424 76489-426 76489-428 76489-430 76489-432 76489-434 76489-436 76489-438 76489-440 76489-442 76489-444 76489-446 76489-448 76489-450 76489-452 76489-454 76489-456 76489-458 76489-460 76489-462 76489-464 76489-466 76489-468 76489-470 76489-472 76489-474 76489-476 76489-478 76489-480 76489-482 76489-484 76489-486 76489-488 76489-490 76489-492 76489-494 76489-496 76489-498 76489-500 76489-502 76489-504
WHEATON® CompletePAK® Sterile Vial Kits, DWK Life Sciences
Vials
CompletePAK® is a range of primary packaging components that are ready-to-use for most drug applications.
  • Off-the-shelf, ready-to-use products
  • One source for total packaging solutions
  • Variety of different sterile components; a kit for any application
  • Complete manufacturing traceability
  • CompletePAK Amber for light-sensitive content packaging
  • CompletePAK ModPAK bulk configurations
Each kit component is particulate cleaned, depyrogenated, and sterilized, arriving in the lab ready-to-use. Vials are packed in a sterile polypropylene tray which can be used as a filling aid. Kit packaging is sized to compliment manual aseptic filling processes.
The CompletePAK® “single source” focus can reduce customer’s supply chain and time to market. Vials, stoppers, and seals are particulate cleaned, depyrogenated and sterilized, arriving in the lab ready-to-use.
Certifications: 1. USP <788> particulate Matter in Injections
2. USP <85> Bacterial Endotoxin Test
3. Lot certified Sterile to SAL <10-6
Delivery information: Batch-specific Certificates of Analysis showing compliance with relevant USP standards are included in each kit.