VWR® SMS Sleeve, Made in USA
Supplier: VWR
VWR®
1024
76491-710CS 202.67 CAD
76491-710
VWR® SMS Sleeve, Made in USA
Disposable Garments
VWR® Made in USA SMS Sleeves are an ideal option for single-use needs.
  • Individually bagged
  • Available with thumbloop configuration
  • Made in USA
A key part of the VWR® Made in USA PPE line, these cleanroom SMS sleeves are 24" in length to provide a comfortable fit while meeting sterility and filtration requirements of many controlled environments.
Packaging: Bagged 1 pair per bag, 100 pairs per case. Packed in poly-lined bag in brown corrugated box.