VWR® Sleeves, Made in USA
Supplier: VWR International
76491-708 76491-714
76491-708CS 484.43 CAD
76491-708 76491-714
VWR® Sleeves, Made in USA
Disposable Garments
VWR® Made in USA Sleeves are an ideal option for single-use needs. Made with the specially formulated Alliance breathable microporous fabric.
  • Individually bagged
  • Available with thumbloop configuration
  • Made with Alliance breathable microporous fabric
  • Made in USA
A key part of the VWR® Made in USA PPE line, these cleanroom sleeves are 21" in length to provide a comfortable fit while meeting sterility and filtration requirements of many controlled environments.
The Alliance fabric is designed to lower particulate levels by providing extremely smooth outer fabric surface that lowers particle cling and build-up in cleanroom applications. Alliance products have a unique antistatic system that assures static dissipation in sensitive cleanroom applications, while providing a liquid-proof barrier that ensures protection from hazardous substances used in Biotech and Pharmaceutical cleanrooms.
Packaging: Bagged 1 pair per bag, 100 pairs per case. Packed in poly-lined bag in brown corrugated box.