VWR® Alliance® Cleanroom Shoe Covers, Anti-Skid
Supplier: VWR International
76491-696 76491-702
76491-696CS 146.97 CAD
76491-696 76491-702
VWR® Alliance® Cleanroom Shoe Covers, Anti-Skid
Disposable Garments
Crafted from durable and breathable Alliance® microporous fabric with a unique anti-skid additive, these shoe covers offer reliable protection and comfort.
  • Ultrasonic welded encapsulated seam construction
  • ¼" top opening width
  • 44 gsm
  • 12" resting length, 6" height, 5" width
  • Non sterile, single-use design
Featuring ultrasonically welded construction, encapsulated elastic, and a flat-welded covered bottom for added comfort, they fit US men's shoe sizes 7 to 13. High-quality and machine-made, all components of these shoe covers are manufactured in the USA.
Packaging:
Single sleeve per bag (poly-lined carton), 150 pieces per case.