Whirl-Pak® Sterilized Filter Bags, Nasco Sampling
Supplier: NASCO SAMPLING LLC
B01385 B01416 B01488 B01348 B01318 B01525
11216-092CS 7819.9 CAD
11216-092 89130-774 89126-754 11216-520 11216-515 75857-054
Whirl-Pak® Sterilized Filter Bags, Nasco Sampling
Bags Blender Bags
Sterilized post manufacturing and specially designed for use with homogenizer blenders. HACCP, EPA, USDA, FDA approved for food contact.
  • Plus 1 sterilization, post-manufacturing sterilization: Increased sample integrity, perforated closure to maintain sterility
  • 330 micron filter layer collecting, blending, pipetting
  • Convenient write-on strip for easy labeling
  • Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring, and closing
  • Opening pull tabs: For easy opening, non-slip textured Tyvek
  • Continuous
More Product Information