Whirl-Pak® Sterilized Thio Bags, Nasco Sampling
Supplier: NASCO SAMPLING LLC
B01403 B01254 B01402 B01040 B01601 B01603
CA11216-780CS 3937.2 CAD
CA11216-780 11216-924 11216-780 11216-922 11216-759 76338-044 76338-046 76338-048
Whirl-Pak® Sterilized Thio Bags, Nasco Sampling
Bags Sample Bags
EPA approved for more sustainable, efficient microbiological water sampling container. A cost effective, low environmental impact alternative to rigid material containers.
  • Plus 1 sterilization, post-manufacturing sterilization: Increased sample integrity, perforated closure to maintain sterility
  • Thiosulfate tablet in the bag, saving technician's time
  • Gusseted bottom option, for self-standing ease
  • Liquid Lock closure for secure samples every time
  • Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring,
More Product Information