Whirl-Pak® Sterilized Write-On Bags, Nasco Sampling
Supplier: NASCO SAMPLING LLC
B01447 B01297 B01062 B01065 B01064 B01445 B01446
CA11216-770CS 3173.4 CAD
CA11216-770 CA11216-779 89130-776 11216-500 11216-770 11216-774 11216-776 11216-772 89132-028 CA11012-434 35000-678
Whirl-Pak® Sterilized Write-On Bags, Nasco Sampling
Bags Sample Bags
Blended, virgin polyethylene film with Write-On strip provides exceptional clarity, superior strength, and sterilized after manufacturing for results you can trust. HACCP, EPA, USDA, FDA approved for food contact.
  • Plus 1 sterilization, post-manufacturing sterilization: Increased sample integrity, perforated closure to maintain sterility
  • Convenient write-on strip for easy labeling
  • Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring, and closing
  • Opening pull tabs: For easy opening, non-slip textured
More Product Information