Oakton® Waterproof PC 450 pH/CON Portable Meters Antylia Scientific

Supplier: Antyila Scientific
35630-10 35630-12 35630-32 35630-80 35630-90
76221-278EA 2801.1 CAD
76221-278 76221-280 76221-282 76221-286 76221-288
Oakton® Waterproof PC 450 pH/CON Portable Meters Antylia Scientific
Electrochemistry Meters pH and Multi-Parameter Meters
More measurements from one meter.

  • Get both pH and conductivity measurements with combination pH/CON probe or separate pH and conductivity probes
  • Use it in any environment with backlit display and rugged design
  • Intuitive, user-friendly icons show electrode status as pH slope and all constant
  • Get quick and easy electrode attachment to a beaker or container with Grip-Clip™ holder
  • Download and analyze up to 500 data sets with USB or RS232 output
  • Use as a benchtop meter as well—built-in stand and wall-mount option

Ideal meter for any environment with itsMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
pH range –2.00...16.00
pH resolution 0.01
pH accuracy ±0.01
pH calibration Up to six points: USA, NIST, DIN, PWB, or custom
Automatic buffer recognition USA, NIST, DIN, PWB, or custom
mV range ±2000 mV
mV resolution 0.1 mV ±999.9 mV; 1 mV beyond
mV accuracy ±0.2 mV ±2 LSD or ±0.5% of reading; whichever is greater
mV calibration ±200 mV
Conductivity range 0 - 200 mS
Conductivity resolution 0.01 µS - 0.1 mS
Conductivity accuracy ±0.5% full-scale (monovalent); ±1% full-scale (divalent)
Salinity range 0 - 50 ppt
Salinity resolution 0.01
Salinity accuracy ±1% full-scale
TDS range 0 - 200 ppt
TDS resolution 0.01 ppm - 0.1 ppt
TDS accuracy ±1% full-scale
TDS calibration Auto/manual; one point per range
Temperature range –10.0…+110.0 °C (–17.0…+230.0 °F)
Temperature resolution 0.1 °F/°C
Temperature accuracy ±0.5 °C (±0.9 °F)
Temperature calibration ±5.0 °C offset
Temperature compensation Automatic or manual from –10 to 110 °C
Display Backlit LCD
Battery life 500 h
Power supply 2 AA batteries
Interfaces RS232 and USB

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR