Oakton® pH 450 Waterproof Portable Meter, Antylia Scientific

Supplier: Antyila Scientific
35618-30 35618-32 35618-90
76221-272EA 2157.82 CAD
76221-272 76221-274 76221-276
Oakton® pH 450 Waterproof Portable Meter, Antylia Scientific
Electrochemistry Meters pH and Multi-Parameter Meters
No more difficulty reading measurements.

  • Use it in any environment with backlit display and rugged design
  • Intuitive, user-friendly icons show electrode status as pH slope
  • Get quick and easy electrode attachment to a beaker or container with Grip-Clip™ holder
  • Download and analyze up to 500 data sets with USB or RS232 output
  • Use as a benchtop meter as well, built-in stand and wall-mount option

Waterproof, IP 67-rated meter is ideal for even the harshest environments. View the main measurement simultaneously with temperature and calibration data with %More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
pH range –2.00...16.00
pH resolution 0.01
pH accuracy ±0.01
pH calibration Up to 6 points: USA, NIST, DIN
Automatic buffer recognition DIN, NIST, USA, and custom
mV range ±2000 mV
mV resolution 0.1 mV ±999.9 mV; 1 mV beyond
mV accuracy ±0.2 mV ±2 LSD or ±0.5% of reading; whichever is greater
mV calibration ±200 mV
ISE range 0.0 - 19,999
ISE resolution 0.01
ISE accuracy ±0.5% full-scale (monovalent); ±1% full-scale (divalent)
ISE calibration Up to 6 points
Temperature range –10.0…+110.0 °C (–17.0…+230.0 °F)
Temperature resolution 0.1 °F/°C
Temperature accuracy ±0.5 °C (±0.9 °F)
Temperature calibration Offset 0.1 increments
Temperature compensation Automatic or manual from –10...110 °C
Display Backlit LCD
Battery life 500 h
Power supply 2 AA batteries
Dimensions 6.5L×3.125W×2.25" H
Interfaces RS232 and USB

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR