Air Cadet® Vacuum/Pressure Pumps, Antylia Scientific
Supplier: Antyila Scientific
07532-40 07532-45 07532-25
89492-594EA 981.54 CAD
89492-594 89492-596 89492-598 89492-602 89492-606
Air Cadet® Vacuum/Pressure Pumps, Antylia Scientific
Pumps Vacuum Pumps
Unique cavity configuration minimizes dead space and air entrapment with unique cavity configuration.
  • No maintenance needed, AC motors are brushless with sealed ball bearings
  • Worry-free pumping
All-plastic, convoluted diaphragm design handles many vacuum/pressure applications and reduces stress, wear, and heat build-up.