Instrument Tray, with Optional Cover, Mortech®
Supplier: Mopec
BA033 BA034 BA035 BA039
76498-206EA 336.22 CAD
76498-206 76498-208 76498-210 76498-218
Instrument Tray, with Optional Cover, Mortech®
Trays/Dishes/Pots Instrument Trays/Dishes
Stainless steel laboratory trays with optional cover.
  • All stainless steel, seamless construction
  • Coved corners for easy cleaning
  • Covers have recessed strap handles for ease in handling and storage
  • Optional cover will fit all three pan sizes
Available in standard or heavyweight construction (please inquire regarding heavyweight construction).
Ordering information: Trays are sold without covers. Covers must be purchased separately.