Borosil® Reusable Class B Serological Pipettes (Pipettes), Foxx Life Sciences®
Supplier: Foxx Life Sciences
7080P01D 7080P02D 7080P05D 7080P06D 7080P09D 7080P11D 7080P22D 7080P41D 7080P42D 7080P55D
76491-368CS 289.87 CAD
76491-368 76491-370 76491-372 76491-374 76491-376 76491-378 76491-380 76491-382 76491-384 76491-386
Borosil® Reusable Class B Serological Pipettes (Pipettes), Foxx Life Sciences®
Pipettes Serological Pipettes
Borosil® brand Pipettes are made of specially-selected, heavy wall, machine-drawn, borosilicate accurate, bore glass tubing. They have precision formed jets, and bevel ground ends that resist chipping.
  • Graduated to tip
  • Two top rings indicate to deliver (TD)
  • Permanent amber stain markings
The graduations are made in durable white enamel or amber stain that is resistant to laboratory reagents and repeat washing. All Pipettes are individually measured and then marked. The uniformity of bore ensures accuracy between any two readings and they areMore Product Information