ECHO Revolve 4, Hybrid Fluorescence Microscope (Upright/Inverted), Discover Echo
Supplier: DISCOVER ECHO INC.
JCND2003 SCND1001 RVSF1000 JCND2005 JCND2004
76299-602EA 3429.18 CAD
76299-602 76299-610 76299-612 76376-718 76376-720 76376-722 76376-724 76376-726 76376-728 76376-730 76376-732 76376-734 76376-736 76490-290 76490-302 76490-308 76490-310 76490-316
ECHO Revolve 4, Hybrid Fluorescence Microscope (Upright/Inverted), Discover Echo
Microscopes
This 'all-in-one' hybrid microscope can be used for upright and inverted observation methods.
  • 5 MP monochrome camera (upgrade from 3.2 MP) - this increases field of view and resolution for fluorescence imaging
  • 5 MP CMOS color camera (brightfield)
  • Intelligent objective nosepiece - this allows the software interface to automatically recognise the objective being used
  • Illumination technique: Brightfield and Epi-fluorescence (phase contrast available as an option)
  • Ultra-bright LEDs with 50000-hour lifespan
  • Touch screen functions drive cameras and illumination,
More Product Information