Cork Dissecting Board with Optional Feet, Mortech®
Supplier: Mopec
BC004 BC005 BC052
76498-290EA 375.6 CAD
76498-290 76498-292 76498-314
Cork Dissecting Board with Optional Feet, Mortech®
Dissecting Boards
Neoprene cork surface offers durability during dissection and is fluid-resistant.
  • 0.25" layer of neoprene cork permanently cemented to a 0.75" marine plywood base
  • Durable and fluid-resistant cork construction
Removable non-skid legs provide an ergonomic, multi-user work surface height.