Minisart® Syringe Filters NML
Supplier: Sartorius
16532GUK 16532K 16532Q 16533GUK 16533K 16533Q 16537K 16537Q 16541K 16541Q 16553K 16569K 16592GUK 16592K 16592Q 16596HNK 16596IGK 1659AHYQ 1751AQ 1757AQ 17593K 17593Q 17594Q 17823K 17823Q 17824K 17824Q 17824R 17825Q 17829K 17829Q S6534FMGUK S6534FMOSK S6534FMQ S6555FMGUK S6555FMOSK S6555FMQ S6596FMOSK S6596FMQ S7594FMOSK S7597FXOSK S7597FXQ S7598FXOSK S7598FXQ
CA76474-362PK 238 CAD
CA76474-362 CA76474-364 CA76474-366 CA76474-368 CA76474-370 CA76474-372 CA76474-374 CA76474-376 CA76474-378 CA76474-380 CA76474-382 CA76474-384 CA76474-386 CA76474-388 CA76474-390 CA76474-392 CA76474-394 CA76474-398 CA76474-402 CA76474-436 CA76474-446 CA76474-448 CA76474-450 CA76474-508 CA76474-510 CA76474-512 CA76474-514 CA76474-516 CA76474-518 CA76474-520 CA76474-522 CA76474-552 CA76474-554 CA76474-556 CA76474-558 CA76474-560 CA76474-562 CA76474-564 CA76474-566 CA76474-570 CA76474-572 CA76474-574 CA76474-576 CA76474-578
Minisart® Syringe Filters NML
Filters Syringe Filters
Minisart NML standard syringe filter for clarification and sterilization of liquids and gases.
  • Largest effective filtration area (EFA) of 6.2 cm²
  • Low absorption
  • High flow rate
  • High total throughput
  • Low hold up volume
  • Gamma-irradiated or EO-sterilized
For clarification and sterilization of liquids, filtration is the optimal method. It removes microorganisms and particles reliably, without any effects on the ingredients due to adsorption or decomposition.
For optimal results, Minisart® Standard syringe filters with an MBS housing provide a choice ofMore Product Information