Borosil® Solution Bottle Borosilicate Glass, Plain Tooled Neck, Foxx Life Sciences®
Supplier: Foxx Life Sciences
BOROSIL®
1585038 1585031 1585046 1585024 1585030 1585033 1585021 1585040 1585029
76490-644EA 455.74 CAD
76490-644 76490-646 76490-634 76490-640 76490-650 76490-642 76490-648 76490-636 76490-638
Borosil® Solution Bottle Borosilicate Glass, Plain Tooled Neck, Foxx Life Sciences®
Bottles Solution Bottles
Borosilicate glass 3.3 bottles, with tooled neck, high mechanical strength and chemical resistant.
  • Bottles are specially designed for handling and storage of culture media
  • Matt-finished enameled area for marking
  • Mechanically strong and chemical resistant
  • Round, no cap
Borosil laboratory bottles are chemically resistant and stable. Fitted with a tooled neck. The bottles are autoclavable and sterilizable.