Borosil® Amber Light-Blocking Reagent Bottles with Ground Glass Stoppers and Graduations, Foxx Life Sciences®
Supplier: Foxx Life Sciences
1509013 1509016 1509017 1509021 1509024 1509029 1509030
76490-620CS 1097.49 CAD
76490-620 76490-622 76490-624 76490-626 76490-628 76490-630 76490-632
Borosil® Amber Light-Blocking Reagent Bottles with Ground Glass Stoppers and Graduations, Foxx Life Sciences®
Bottles Glass Bottles
Amber borosilicate glass 3.3 bottles, with flat head solid glass stoppers.
  • Narrow mouth with ground neck
  • Matt-Finished enameled area for marking
  • Mechanically strong and chemical resistant
  • Supplied with solid glass stopper
  • Graduated
  • Amber
Certifications: Complies with IS 1388 (Part II), EN iso 4796-2.