Borosil® Reagent Bottles Plain Narrow Mouth, Graduated with Interchangable Glass Stopper, Foxx Life Sciences®
Supplier: Foxx Life Sciences
BOROSIL®
1500016 1500029 1500017 1500013 1500030 1500021 1500024
76490-600CS 911.33 CAD
76490-600 76490-602 76490-596 76490-598 76490-592 76490-594 76490-590
Borosil® Reagent Bottles Plain Narrow Mouth, Graduated with Interchangable Glass Stopper, Foxx Life Sciences®
Bottles Glass Bottles
Borosilicate glass 3.3 bottles, with flat head solid glass stoppers.
  • Narrow mouth with ground neck
  • Matt-Finished enameled area for marking
  • Mechanically strong and chemical resistant
  • Supplied with solid glass stopper
  • Graduated
Borosil® laboratory bottles are chemically resistant and stable. Supplied with a interchangable Glass stopper and are graduated. The Tottles and Stoppers caps are autoclavable and sterilizable.
Certifications: Complies with IS 1388 (Part II), EN ISO 4796-2.